Product Details

SNP ID
rs199841978
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:46688643 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGAGGTGCACGGGTGCGTCCTGG[A/G]CGTGCTGCTGCTCCATCGCCTGGTC
Phenotype
MIM: 611200
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101926934 PubMed Links

Gene Details

Gene
LOC101926934
Gene Name
uncharacterized LOC101926934
There are no transcripts associated with this gene.

Gene
TDRD6
Gene Name
tudor domain containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010870.2 769 Missense Mutation GAC,GGC D172G NP_001010870.1
NM_001168359.1 769 Missense Mutation GAC,GGC D172G NP_001161831.1

View Full Product Details