Product Details

SNP ID
rs199528335
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:24650843 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCCAGTGATCACTAGGAAATCTA[C/T]CACAGTCCAGTTTTTCTAATCCAAG
Phenotype
MIM: 609269 MIM: 605764
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIAA0319 PubMed Links

Gene Details

Gene
KIAA0319
Gene Name
KIAA0319
There are no transcripts associated with this gene.

Gene
TDP2
Gene Name
tyrosyl-DNA phosphodiesterase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016614.2 1059 Missense Mutation GAT,GGT D345G NP_057698.2

View Full Product Details