Product Details

SNP ID
rs200979746
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:36155065 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGATTATCTCCACTCGGCCGTC[C/T]GTGGAGGCCTTTGGAGGCACAAGGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EEPD1 PubMed Links

Gene Details

Gene
EEPD1
Gene Name
endonuclease/exonuclease/phosphatase family domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030636.2 1459 Silent Mutation TCC,TCT S247S NP_085139.2
XM_017012657.1 1459 Silent Mutation TCC,TCT S247S XP_016868146.1
XM_017012658.1 1459 Silent Mutation TCC,TCT S247S XP_016868147.1

View Full Product Details