Product Details
- SNP ID
-
rs201069425
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:100721963 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCCCCGGGCTGTGTGCATTTCAGA[C/T]GGGCTGTGCTGAACACTGCAGCTTG
- Phenotype
-
MIM: 133170
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
EPO
PubMed Links
Gene Details
- Gene
- EPO
- Gene Name
- erythropoietin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000799.2 |
342 |
Missense Mutation |
ACG,ATG |
T54M |
NP_000790.2 |
View Full Product Details