Product Details

SNP ID
rs201072665
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:120275302 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTAAAGAACTGCCCTGTGGAGAG[A/C]GGTATGCTGTGGCCTTCTTCTGCTT
Phenotype
MIM: 605410
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KCND2 PubMed Links

Gene Details

Gene
KCND2
Gene Name
potassium voltage-gated channel subfamily D member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012281.2 1635 Silent Mutation AGG,CGG R224R NP_036413.1
XM_011516165.2 1635 Silent Mutation AGG,CGG R224R XP_011514467.1

View Full Product Details