Product Details
- SNP ID
-
rs200800005
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:100182149 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GACTTTGAAGACAGCTTGAATCGCA[A/G]TGTGAAGAAGAGAGCAGCAAAACGA
- Phenotype
-
MIM: 608489
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GATS
PubMed Links
Gene Details
- Gene
- GATS
- Gene Name
- GATS, stromal antigen 3 opposite strand
There are no transcripts associated with this gene.
- Gene
- GPC2
- Gene Name
- glypican 2
There are no transcripts associated with this gene.
- Gene
- STAG3
- Gene Name
- stromal antigen 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001282716.1 |
470 |
Missense Mutation |
AAT,AGT |
N59S |
NP_001269645.1 |
NM_001282717.1 |
470 |
Missense Mutation |
AAT,AGT |
N59S |
NP_001269646.1 |
NM_001282718.1 |
470 |
Missense Mutation |
AAT,AGT |
N59S |
NP_001269647.1 |
NM_012447.3 |
470 |
Missense Mutation |
AAT,AGT |
N59S |
NP_036579.2 |
XM_011515742.1 |
470 |
Missense Mutation |
AAT,AGT |
N59S |
XP_011514044.1 |
XM_017011683.1 |
470 |
Missense Mutation |
AAT,AGT |
N59S |
XP_016867172.1 |
XM_017011684.1 |
470 |
Missense Mutation |
AAT,AGT |
N59S |
XP_016867173.1 |
XM_017011685.1 |
470 |
Missense Mutation |
AAT,AGT |
N59S |
XP_016867174.1 |
XM_017011686.1 |
470 |
Missense Mutation |
AAT,AGT |
N59S |
XP_016867175.1 |
XM_017011687.1 |
470 |
Missense Mutation |
AAT,AGT |
N59S |
XP_016867176.1 |
View Full Product Details