Product Details

SNP ID
rs200517168
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:77167765 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATGCCATAGTGCCCACAATTAAC[A/G]TTCAAGATGAGCGGTTTGTTGATTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC146 PubMed Links

Gene Details

Gene
CCDC146
Gene Name
coiled-coil domain containing 146
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020879.2 224 Missense Mutation ATT,GTT I33V NP_065930.2

View Full Product Details