Product Details

SNP ID
rs200989750
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:36396278 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTATTTATGTAGAAACTGCTGGAG[C/T]GAACCCGTGCCAGGCGAGAGAATCT
Phenotype
MIM: 616027
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANLN PubMed Links

Gene Details

Gene
ANLN
Gene Name
anillin actin binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284301.2 252 Nonsense Mutation CGA,TGA R11* NP_001271230.1
NM_001284302.2 252 Nonsense Mutation CGA,TGA R11* NP_001271231.1
NM_018685.4 252 Nonsense Mutation CGA,TGA R11* NP_061155.2
XM_006715746.1 252 Nonsense Mutation CGA,TGA R11* XP_006715809.1
XM_006715747.3 252 Nonsense Mutation CGA,TGA R11* XP_006715810.1
XM_017012354.1 252 Nonsense Mutation CGA,TGA R11* XP_016867843.1
XM_017012355.1 252 Nonsense Mutation CGA,TGA R11* XP_016867844.1
XM_017012356.1 252 Nonsense Mutation CGA,TGA R11* XP_016867845.1
Gene
KIAA0895
Gene Name
KIAA0895
There are no transcripts associated with this gene.

View Full Product Details