Product Details
- SNP ID
-
rs199957832
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:2538468 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGTGGCAGAGAGCCCTCAGTGCCTC[A/G]GTGAGTGGCTGGGCTGGGGCCACCT
- Phenotype
-
MIM: 614506
MIM: 602576
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
BRAT1
PubMed Links
Gene Details
- Gene
- BRAT1
- Gene Name
- BRCA1 associated ATM activator 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_152743.3 |
2298 |
Silent Mutation |
ACC,ACT |
T689T |
NP_689956.2 |
XM_005249643.4 |
2298 |
Silent Mutation |
ACC,ACT |
T749T |
XP_005249700.1 |
XM_011515177.2 |
2298 |
Silent Mutation |
ACC,ACT |
T777T |
XP_011513479.1 |
XM_011515178.1 |
2298 |
Silent Mutation |
ACC,ACT |
T777T |
XP_011513480.1 |
XM_011515179.2 |
2298 |
Silent Mutation |
ACC,ACT |
T776T |
XP_011513481.1 |
XM_011515181.2 |
2298 |
Silent Mutation |
ACC,ACT |
T717T |
XP_011513483.1 |
XM_011515182.2 |
2298 |
Silent Mutation |
ACC,ACT |
T777T |
XP_011513484.1 |
XM_011515183.1 |
2298 |
Silent Mutation |
ACC,ACT |
T602T |
XP_011513485.1 |
XM_011515184.2 |
2298 |
Silent Mutation |
ACC,ACT |
T602T |
XP_011513486.1 |
XM_011515186.2 |
2298 |
UTR 3 |
|
|
XP_011513488.1 |
XM_017011833.1 |
2298 |
Silent Mutation |
ACC,ACT |
T748T |
XP_016867322.1 |
XM_017011834.1 |
2298 |
Silent Mutation |
ACC,ACT |
T688T |
XP_016867323.1 |
XM_017011835.1 |
2298 |
Silent Mutation |
ACC,ACT |
T514T |
XP_016867324.1 |
XM_017011836.1 |
2298 |
UTR 3 |
|
|
XP_016867325.1 |
- Gene
- LFNG
- Gene Name
- LFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase
There are no transcripts associated with this gene.
View Full Product Details