Product Details

SNP ID
rs200714078
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:7639133 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCTGAAAGAATAAACATTTTTCC[A/G]GTGGGATGAATCTAAAAACGAAACA
Phenotype
MIM: 179837
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RPA3 PubMed Links

Gene Details

Gene
RPA3
Gene Name
replication protein A3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002947.4 1283 Silent Mutation NP_002938.1
Gene
UMAD1
Gene Name
UBAP1-MVB12-associated (UMA) domain containing 1
There are no transcripts associated with this gene.

View Full Product Details