Product Details

SNP ID
rs200866893
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:146116949 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATTGTCAGCAGCTGCCTCTGCAGA[A/G]CCTGGACGGCTCCCTCCACGTCCCG
Phenotype
MIM: 604569
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CNTNAP2 PubMed Links

Gene Details

Gene
CNTNAP2
Gene Name
contactin associated protein-like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014141.5 915 Missense Mutation ACC,GCC T25A NP_054860.1
XM_017011950.1 915 Missense Mutation ACC,GCC T25A XP_016867439.1

View Full Product Details