Product Details

SNP ID
rs201712782
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:99333415 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCATGCCTGGAACAGGGATCGTA[C/T]TCGTAAGTATTTTATTAACTTTGCT
Phenotype
MIM: 604220
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARPC1A PubMed Links

Gene Details

Gene
ARPC1A
Gene Name
actin related protein 2/3 complex subunit 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190996.1 Intron NP_001177925.1
NM_006409.3 Intron NP_006400.2

View Full Product Details