Product Details

SNP ID
rs201434174
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:116500388 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAGTTCCTGACGGTGTTCCTGGC[C/T]ATTCCCCTGGCCTTCATTGCGGGAA
Phenotype
MIM: 601048
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CAV2 PubMed Links

Gene Details

Gene
CAV2
Gene Name
caveolin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206747.1 460 Silent Mutation GCC,GCT A80A NP_001193676.1
NM_001206748.1 460 Silent Mutation GCC,GCT A10A NP_001193677.1
NM_001233.4 460 Silent Mutation GCC,GCT A93A NP_001224.1
NM_198212.2 460 Intron NP_937855.1

View Full Product Details