Product Details

SNP ID
rs201118795
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:149004029 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTTGGCCCTTGTACTTCTTGGCC[A/G]GGCTGTTGTACACGGGCTCTAGCTG
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
PDIA4 PubMed Links

Gene Details

Gene
PDIA4
Gene Name
protein disulfide isomerase family A member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004911.4 1985 Missense Mutation CCG,CTG P568L NP_004902.1
XM_006716185.2 1985 Missense Mutation CCG,CTG P569L XP_006716248.1

View Full Product Details