Product Details
- SNP ID
-
hCV191840030
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:75813361 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGGACAGCTGGTAGCTGACCACTC[A/G]GTTCTCAGGAATTCTCTTGGAAACA
- Phenotype
-
MIM: 602495
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CCL24
PubMed Links
Gene Details
- Gene
- CCL24
- Gene Name
- C-C motif chemokine ligand 24
There are no transcripts associated with this gene.
View Full Product Details