Product Details

SNP ID
rs201608824
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:156950209 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGGCCAAGGCCACGGCCGCCACC[C/G]CAAAGACCAGACCCTCCGCAGCCGC
Phenotype
MIM: 611269
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NOM1 PubMed Links

Gene Details

Gene
NOM1
Gene Name
nucleolar protein with MIF4G domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138400.1 487 Missense Mutation CCA,GCA P158A NP_612409.1

View Full Product Details