Product Details

SNP ID
rs201126235
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:99892516 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTCCAGACTATCTCTACTCTCAA[C/G]TTCCCAGTAATGTTTCCCTGAGGTG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TRIM4 PubMed Links

Gene Details

Gene
TRIM4
Gene Name
tripartite motif containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033017.3 1271 Missense Mutation CTT,GTT L384V NP_148977.2
NM_033091.2 1271 Missense Mutation CTT,GTT L358V NP_149082.1

View Full Product Details