Product Details

SNP ID
rs201481844
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:44965718 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCCACACAAGGTCACGCCCGTAG[A/G]GTGGCGGCTGCCTTGCAGCCTGGAA
Phenotype
MIM: 600642
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MYO1G PubMed Links

Gene Details

Gene
MYO1G
Gene Name
myosin IG
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033054.2 2203 Missense Mutation CCC,CTC P767L NP_149043.2
XM_017012503.1 2203 Missense Mutation CCC,CTC P652L XP_016867992.1
XM_017012504.1 2203 Missense Mutation CCC,CTC P529L XP_016867993.1

View Full Product Details