Product Details

SNP ID
rs199575787
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:99876202 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATTCAATGTGGGCGTAATGATGC[G/T]GTAGAACACAGAACCAACTTTGTTC
Phenotype
MIM: 606534
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CYP3A43 PubMed Links

Gene Details

Gene
CYP3A43
Gene Name
cytochrome P450 family 3 subfamily A member 43
There are no transcripts associated with this gene.

Gene
OR2AE1
Gene Name
olfactory receptor family 2 subfamily AE member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005276.1 832 Missense Mutation AGC,CGC S278R NP_001005276.1

View Full Product Details