Product Details

SNP ID
rs199696106
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:23317681 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGTCCAGTGATAATCACCATCCT[A/C]ACTTTAGCATCTGGTGCTTCCGCTG
Phenotype
MIM: 608259 MIM: 614624
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
IGF2BP3 PubMed Links

Gene Details

Gene
IGF2BP3
Gene Name
insulin like growth factor 2 mRNA binding protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006547.2 1932 Intron NP_006538.2
XM_006715639.2 1932 Intron XP_006715702.1
XM_011515089.2 1932 Silent Mutation GTG,GTT V456V XP_011513391.1
XM_011515090.2 1932 Silent Mutation GTG,GTT V343V XP_011513392.1
XM_011515091.2 1932 Intron XP_011513393.1
XM_011515092.2 1932 Intron XP_011513394.1
XM_011515093.2 1932 Silent Mutation GTG,GTT V358V XP_011513395.1
Gene
MALSU1
Gene Name
mitochondrial assembly of ribosomal large subunit 1
There are no transcripts associated with this gene.

View Full Product Details