Product Details

SNP ID
rs201252065
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:94832133 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTAAATGGAGAAACTGAGAGTTCT[A/G]CTGCTGGATTGAAAGTCAAAACCGA
Phenotype
MIM: 611351
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
INTS8 PubMed Links

Gene Details

Gene
INTS8
Gene Name
integrator complex subunit 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017864.3 854 Missense Mutation ACT,GCT T238A NP_060334.2
XM_017013616.1 854 Missense Mutation ACT,GCT T238A XP_016869105.1
XM_017013617.1 854 Missense Mutation ACT,GCT T238A XP_016869106.1
XM_017013618.1 854 Missense Mutation ACT,GCT T129A XP_016869107.1
XM_017013619.1 854 UTR 5 XP_016869108.1

View Full Product Details