Product Details

SNP ID
rs201444798
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:23298877 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCCAGATGCGGTGGCCGTCATAG[C/T]GGCTCCTGCATTTCATGATGTTGTT
Phenotype
MIM: 606663
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOXL2 PubMed Links

Gene Details

Gene
LOXL2
Gene Name
lysyl oxidase like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002318.2 2543 Missense Mutation CAC,CGC H735R NP_002309.1
Gene
R3HCC1
Gene Name
R3H domain and coiled-coil containing 1
There are no transcripts associated with this gene.

View Full Product Details