Product Details

SNP ID
rs202025915
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:81443444 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGCACACCACGCCCTTCATTTTA[C/T]ATTCCTTAAAAAAGAGAGAGGTTAA
Phenotype
MIM: 170715
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PMP2 PubMed Links

Gene Details

Gene
PMP2
Gene Name
peripheral myelin protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002677.3 451 Missense Mutation NP_002668.1

View Full Product Details