Product Details
- SNP ID
-
rs199993571
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:70058650 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTCCTAATACTCACCTTGTGAGGC[C/T]GGTGCTTCTCATGAACATGAAGAAT
- Phenotype
-
MIM: 611781
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PRDM14
PubMed Links
Gene Details
- Gene
- PRDM14
- Gene Name
- PR domain 14
View Full Product Details