Product Details

SNP ID
rs200016935
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:123775281 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGAGGATAACACCATTTTCATAT[C/T]ATACTGGGATTATGGAGGTGAGTTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM91A1 PubMed Links

Gene Details

Gene
FAM91A1
Gene Name
family with sequence similarity 91 member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317917.1 556 UTR 5 NP_001304846.1
NM_001317918.1 556 Missense Mutation CAT,TAT H98Y NP_001304847.1
NM_144963.3 556 Missense Mutation CAT,TAT H98Y NP_659400.2

View Full Product Details