Product Details

SNP ID
rs200491543
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.8:104491003 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTCAAAGGACTCTGGTTCTGACT[C/T]AGGGAACTTGATCGTCCTAATGTAA
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
LRP12 PubMed Links

Gene Details

Gene
LRP12
Gene Name
LDL receptor related protein 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135703.2 2377 Silent Mutation CTA,CTG L731L NP_001129175.1
NM_013437.4 2377 Silent Mutation CTA,CTG L750L NP_038465.1
Gene
MIR548A3
Gene Name
microRNA 548a-3
There are no transcripts associated with this gene.

View Full Product Details