Product Details

SNP ID
rs200661557
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:93922569 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATTGCTGTCTCTTTGTTACCCCA[A/T]GAGACTTTGCTAGAGATTGAAAATG
Phenotype
MIM: 605993
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MIR378D2 PubMed Links

Gene Details

Gene
MIR378D2
Gene Name
microRNA 378d-2
There are no transcripts associated with this gene.

Gene
PDP1
Gene Name
pyruvate dehyrogenase phosphatase catalytic subunit 1
There are no transcripts associated with this gene.

View Full Product Details