Product Details

SNP ID
rs200323201
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.8:39584646 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATGTTCCTTCTCCTCGCCCTCCT[A/C]ACTGAGCTTGGAAGACTGCAAGCCC
Phenotype
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
ADAM18 PubMed Links

Gene Details

Gene
ADAM18
Gene Name
ADAM metallopeptidase domain 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190956.1 79 Silent Mutation CTA,CTC L8L NP_001177885.1
NM_001320313.1 79 Silent Mutation CTA,CTC L8L NP_001307242.1
NM_014237.2 79 Silent Mutation CTA,CTC L8L NP_055052.1
Gene
LOC100130964
Gene Name
ADAM metallopeptidase domain 3A-like
There are no transcripts associated with this gene.

View Full Product Details