Product Details

SNP ID
rs200494214
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:144336084 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCGTGCAGTGGCGCGCCGAGGTCG[C/T]TGCGGGCGCGTCCGTAGGCGCTCTC
Phenotype
MIM: 604900 MIM: 605858
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DGAT1 PubMed Links

Gene Details

Gene
DGAT1
Gene Name
diacylglycerol O-acyltransferase 1
There are no transcripts associated with this gene.

Gene
SCRT1
Gene Name
scratch family transcriptional repressor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031309.5 198 Missense Mutation AAC,AGC N29S NP_112599.2

View Full Product Details