Product Details
- SNP ID
-
rs200626858
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:71843762 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTCCAGCTTGGAGAGCTTAGTGTC[G/T]GGGGGCACCCAGGGCAGGCTGGTCT
- Phenotype
-
MIM: 603628
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MSC
PubMed Links
Gene Details
- Gene
- MSC
- Gene Name
- musculin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005098.3 |
735 |
Silent Mutation |
|
|
NP_005089.2 |
- Gene
- MSC-AS1
- Gene Name
- MSC antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details