Product Details

SNP ID
rs201417292
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:19049836 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATTCCTAGGGAACCTGGTGCTGAA[C/T]CTGTGGGACTGTGGCGGTCAGGACA
Phenotype
MIM: 613433 MIM: 612194 MIM: 616292
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HAUS6 PubMed Links

Gene Details

Gene
HAUS6
Gene Name
HAUS augmin like complex subunit 6
There are no transcripts associated with this gene.

Gene
RRAGA
Gene Name
Ras related GTP binding A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006570.4 463 Silent Mutation AAC,AAT N59N NP_006561.1
Gene
SAXO1
Gene Name
stabilizer of axonemal microtubules 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287049.1 463 Intron NP_001273978.1
NM_001287050.1 463 Intron NP_001273979.1
NM_153707.3 463 Intron NP_714918.2
XM_011517744.1 463 Intron XP_011516046.1
XM_011517745.1 463 Intron XP_011516047.1
XM_011517746.2 463 Intron XP_011516048.1
XM_011517747.2 463 Intron XP_011516049.1
XM_017014314.1 463 Intron XP_016869803.1
XM_017014315.1 463 Intron XP_016869804.1

View Full Product Details