Product Details

SNP ID
rs200939909
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:42187124 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTCCAGGCCCAGCCCCTGTCCCA[T/C]CGCCAACCCTTTATTTCATCCACAC
Phenotype
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
SPATA31A6 PubMed Links

Gene Details

Gene
SPATA31A6
Gene Name
SPATA31 subfamily A member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145196.1 Intron NP_001138668.1
XM_011517871.2 Intron XP_011516173.1

View Full Product Details