Product Details
- SNP ID
-
rs200409116
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:99915271 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAAGATGAAGAAAAAGTGAAATTAC[A/G]CCGTCTTGAACCAGCTATCCAGAAA
- Phenotype
-
MIM: 604204
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
STX17
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs41305451] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- STX17
- Gene Name
- syntaxin 17
- Gene
- STX17-AS1
- Gene Name
- STX17 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details