Product Details

SNP ID
rs201397337
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:35957627 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAATTGCCAAGGATGTGTGTACCA[C/T]GGAAGCAGCACCACCAATAGCCCAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR2S2 PubMed Links

Gene Details

Gene
OR2S2
Gene Name
olfactory receptor family 2 subfamily S member 2 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019897.2 528 Missense Mutation ATG,GTG M158V NP_063950.2

View Full Product Details