Product Details

SNP ID
rs200288462
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:136673857 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATGGCCCGGTGGCAGGTGTCCAC[A/G]AGCGCAGGGACGTCCGCCGCAGTGA
Phenotype
MIM: 603100 MIM: 608582 MIM: 611767
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AGPAT2 PubMed Links

Gene Details

Gene
AGPAT2
Gene Name
1-acylglycerol-3-phosphate O-acyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012727.1 738 Silent Mutation CTC,CTT L212L NP_001012745.1
NM_006412.3 738 Silent Mutation CTC,CTT L244L NP_006403.2
Gene
EGFL7
Gene Name
EGF like domain multiple 7
There are no transcripts associated with this gene.

Gene
MIR126
Gene Name
microRNA 126
There are no transcripts associated with this gene.

View Full Product Details