Product Details

SNP ID
rs200847005
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:128721398 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGTAGAGGTGCGAGACGAGGAGC[A/G]GGCTGGCCACCAACGAGAAGAGGGA
Phenotype
MIM: 610714
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC100506100 PubMed Links

Gene Details

Gene
LOC100506100
Gene Name
uncharacterized LOC100506100
There are no transcripts associated with this gene.

Gene
PKN3
Gene Name
protein kinase N3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317926.1 775 Intron NP_001304855.1
NM_013355.4 775 Intron NP_037487.2
XM_005251946.3 775 Intron XP_005252003.1
XM_006717080.2 775 Intron XP_006717143.1
XM_017014649.1 775 Intron XP_016870138.1
XM_017014650.1 775 Intron XP_016870139.1
Gene
ZDHHC12
Gene Name
zinc finger DHHC-type containing 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318015.1 775 Missense Mutation CCG,CTG P251L NP_001304944.1
NM_001318016.1 775 Silent Mutation CCC,CCT P205P NP_001304945.1
NM_001318020.1 775 Missense Mutation CGC,TGC R189C NP_001304949.1
NM_001318023.1 775 Missense Mutation CCG,CTG P195L NP_001304952.1
NM_032799.4 775 Missense Mutation CCG,CTG P196L NP_116188.2
XM_011519116.2 775 Missense Mutation CGC,TGC R244C XP_011517418.1
Gene
ZER1
Gene Name
zyg-11 related cell cycle regulator
There are no transcripts associated with this gene.

View Full Product Details