Product Details

SNP ID
rs201195349
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:77728528 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTGACTTACCATCATATTCTGGG[A/G]AGTAGTCGACTAGATGGGAATACAT
Phenotype
MIM: 600998
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GNAQ PubMed Links

Gene Details

Gene
GNAQ
Gene Name
G protein subunit alpha q
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002072.4 1089 Missense Mutation TCC,TTC S292F NP_002063.2
XM_017014628.1 1089 Missense Mutation TCC,TTC S234F XP_016870117.1

View Full Product Details