Product Details

SNP ID
rs200354889
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:6328684 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGATCTGACAGAGCCCGAGCAAA[C/G]AGAGCTCAAAACCAAACTCACTAAA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TPD52L3 PubMed Links

Gene Details

Gene
TPD52L3
Gene Name
tumor protein D52-like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001874.2 336 Missense Mutation ACA,AGA T30R NP_001001874.1
NM_001001875.3 336 Missense Mutation ACA,AGA T30R NP_001001875.1
NM_033516.5 336 Missense Mutation ACA,AGA T30R NP_277051.3
XM_017015280.1 336 Missense Mutation ACA,AGA T30R XP_016870769.1

View Full Product Details