Product Details

SNP ID
rs200561286
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:124786983 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGCCCCCCGCAACAGGTGTTTGG[A/G]GACCTGGACCAGGTGAGGATGACCT
Phenotype
MIM: 615899
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OLFML2A PubMed Links

Gene Details

Gene
OLFML2A
Gene Name
olfactomedin like 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282715.1 212 Intron NP_001269644.1
NM_182487.3 212 Silent Mutation GGA,GGG G33G NP_872293.2
XM_005251760.4 212 Silent Mutation GGA,GGG G33G XP_005251817.1
XM_006716989.2 212 Silent Mutation GGA,GGG G33G XP_006717052.1

View Full Product Details