Product Details

SNP ID
rs200585902
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:124787176 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTACCGCACCTCCCTCCTCTCTC[A/G]ACCCCTGTGAGAACGAGTGGAAGAT
Phenotype
MIM: 615899
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OLFML2A PubMed Links

Gene Details

Gene
OLFML2A
Gene Name
olfactomedin like 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282715.1 405 Intron NP_001269644.1
NM_182487.3 405 Missense Mutation AAC,GAC N98D NP_872293.2
XM_005251760.4 405 Missense Mutation AAC,GAC N98D XP_005251817.1
XM_006716989.2 405 Missense Mutation AAC,GAC N98D XP_006717052.1

View Full Product Details