Product Details

SNP ID
rs200933289
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:1387311 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCCCGGACTGCATCATCATGCGC[C/T]GCCGCACCGTGTCGAAGGGGTAGGA
Phenotype
MIM: 308385 MIM: 430000
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IL3RA PubMed Links

Gene Details

Gene
IL3RA
Gene Name
interleukin 3 receptor subunit alpha
There are no transcripts associated with this gene.

View Full Product Details