Product Details

SNP ID
rs199894106
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:69013117 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTCCTCGTAGTCGTCAAAGTTAC[A/T]GGCATCCCCAGGGCCTGTGTACTTC
Phenotype
MIM: 602745 MIM: 176893
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PIP5K1B PubMed Links

Gene Details

Gene
PIP5K1B
Gene Name
phosphatidylinositol-4-phosphate 5-kinase type 1 beta
There are no transcripts associated with this gene.

Gene
PRKACG
Gene Name
protein kinase cAMP-activated catalytic subunit gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002732.3 1007 Missense Mutation AGT,TGT S326C NP_002723.2

View Full Product Details