Product Details

SNP ID
rs200081386
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:122789249 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACACAGCCTGTGCCCTCCAGATGT[C/T]TGTCTTTGCAGGTCTGGCTGATACT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR5C1 PubMed Links

Gene Details

Gene
OR5C1
Gene Name
olfactory receptor family 5 subfamily C member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001923.1 317 Missense Mutation TCT,TTT S106F NP_001001923.1

View Full Product Details