Product Details

SNP ID
rs200117818
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:34179039 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGAGTGGGGCGGTGAGGGGAAGG[A/T]GGAGGGAAGTAGGACTTCAACATGG
Phenotype
MIM: 609787
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC114224 PubMed Links

Gene Details

Gene
LOC114224
Gene Name
uncharacterized LOC114224
There are no transcripts associated with this gene.

Gene
UBAP1
Gene Name
ubiquitin associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171201.1 35 Missense Mutation AGG,TGG R9W NP_001164672.1
NM_001171202.1 35 Missense Mutation AGG,TGG R9W NP_001164673.1
NM_001171203.2 35 UTR 5 NP_001164674.1
NM_001171204.2 35 UTR 5 NP_001164675.1
NM_016525.4 35 UTR 5 NP_057609.2
XM_006716779.3 35 Intron XP_006716842.1
XM_011517898.2 35 Intron XP_011516200.1
XM_011517899.1 35 Intron XP_011516201.1
XM_011517900.1 35 Intron XP_011516202.1
XM_017014801.1 35 Intron XP_016870290.1
XM_017014802.1 35 Intron XP_016870291.1

View Full Product Details