Product Details

SNP ID
rs199518594
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:129321865 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTCACCTGTCGGGGGAGGTGGGC[A/G]AGGAAGCCTGCAGCTCCTGGAACCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C9orf106 PubMed Links

Gene Details

Gene
C9orf106
Gene Name
chromosome 9 open reading frame 106
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012715.3 105 Missense Mutation AAG,GAG K18E NP_001012733.2

View Full Product Details