Product Details

SNP ID
rs202221094
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:22006082 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAGACGACCCCAGGCATCGCGCA[C/T]GTCCAGCCGCGCCCCGGCCCGGTGC
Phenotype
MIM: 600431 MIM: 613149
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDKN2B PubMed Links

Gene Details

Gene
CDKN2B
Gene Name
cyclin dependent kinase inhibitor 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004936.3 805 Missense Mutation ATG,GTG M108V NP_004927.2
NM_078487.2 805 UTR 3 NP_511042.1
Gene
CDKN2B-AS1
Gene Name
CDKN2B antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details