Product Details

SNP ID
rs201954758
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:19050358 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCACCGGTTTGAGAAGATCAGCAA[C/T]ATCATCAAACAGTTCAAGCTGAGCT
Phenotype
MIM: 613433 MIM: 612194 MIM: 616292
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HAUS6 PubMed Links

Gene Details

Gene
HAUS6
Gene Name
HAUS augmin like complex subunit 6
There are no transcripts associated with this gene.

Gene
RRAGA
Gene Name
Ras related GTP binding A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006570.4 985 Silent Mutation AAC,AAT N233N NP_006561.1
Gene
SAXO1
Gene Name
stabilizer of axonemal microtubules 1
There are no transcripts associated with this gene.

View Full Product Details