Product Details

SNP ID
rs202119401
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:34993271 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTACAAGATTCTTGGGATCCCAT[C/T]GGGGGCCAACGAGGATGAGATCAAG
Phenotype
MIM: 611328
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNAJB5 PubMed Links

Gene Details

Gene
DNAJB5
Gene Name
DnaJ heat shock protein family (Hsp40) member B5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135004.2 182 Missense Mutation TCG,TTG S127L NP_001128476.2
NM_001135005.2 182 Missense Mutation TCG,TTG S85L NP_001128477.1
NM_012266.5 182 Missense Mutation TCG,TTG S13L NP_036398.3
XM_005251428.3 182 Missense Mutation TCG,TTG S13L XP_005251485.1
XM_006716751.2 182 Missense Mutation TCG,TTG S13L XP_006716814.1
XM_011517842.2 182 Missense Mutation TCG,TTG S85L XP_011516144.1
Gene
DNAJB5-AS1
Gene Name
DNAJB5 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details