Product Details
- SNP ID
-
rs202000235
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:6012850 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCAGGGTTGTTATCTCGCATATTTA[A/G]TAGCATTGGCCACCGAAAATATTCT
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
KIAA2026
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1411949] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KIAA2026
- Gene Name
- KIAA2026
There are no transcripts associated with this gene.
- Gene
- MIR4665
- Gene Name
- microRNA 4665
There are no transcripts associated with this gene.
- Gene
- RANBP6
- Gene Name
- RAN binding protein 6
View Full Product Details