Product Details

SNP ID
rs201589514
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:124787022 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGGATGACCTCGGAGGGCTCCGA[C/G]TGCCGTTGCAAGTGCATCATGCGGC
Phenotype
MIM: 615899
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OLFML2A PubMed Links

Gene Details

Gene
OLFML2A
Gene Name
olfactomedin like 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282715.1 251 Intron NP_001269644.1
NM_182487.3 251 Missense Mutation GAC,GAG D46E NP_872293.2
XM_005251760.4 251 Missense Mutation GAC,GAG D46E XP_005251817.1
XM_006716989.2 251 Missense Mutation GAC,GAG D46E XP_006717052.1

View Full Product Details